Quick-DNA™ Plus Kits, Zymo Research
Supplier: Zymo Research
DNA from any sample (cells, tissues, biological fluids and more)
- Quick and easy: Simple 20 minute procedure
- Highest yield: Recover 3× more DNA
- Ultra-pure: Ready for qPCR, next-generation sequencing, arrays and more
The Quick-DNA™ plus kits are the easiest method for high yield total DNA extraction (e.g., genomic, mitochondrial, viral) from any biological fluid, cell culture, or solid tissue sample. Innovative reagents and zymo-spin technology allow for ultra-pure and concentrated genomic DNA ˃50 kb to be eluted in as little as 10 µl. Zymo-Spin™ columns ensure no buffer retention. Purified DNA is RNA-free bypassing the need for RNase A treatment and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays and methylation analysis.