Volumetric Flasks, Class A, Wide Mouth, Heavy Duty, with PE Stopper, Chemglass
Supplier: Chemglass
CG-1619-1L CG-1619-2L CG-1619-05 CG-1619-10 CG-1619-20 CG-1619-25 CG-1619-50 CG-1619-100 CG-1619-200 CG-1619-250 CG-1619-500
89038-420EA 723 CAD
89038-420 89038-422 89038-402 89038-404 89038-406 89038-408 89038-410 89038-412 89038-414 89038-416 89038-418
Volumetric Flasks, Class A, Wide Mouth, Heavy Duty, with PE Stopper, Chemglass
Flasks Volumetric Flasks
These volumetric flasks with wide mouth design facilitate mixing, pipette access, sampling and filling.
  • Clearly visible large permanent white letters
  • Heavy duty design
The unique flat bottom interior permits the use of a standard stir bar.
Certifications: Flasks are calibrated "To Contain" at Class A tolerances, per ASTM E288 specifications. Volumetric flask capacities of 20 ml, 500 ml, 1000 ml and 2000 ml are not included in the ASTM E288 specifications; however, flasks are calibrated to Class A tolerances.
Ordering information: Supplied complete with PE stopper.