Boreal C-SCOPE Compound Microscopes
Supplier: VWR International
1100100403313 1100100403352
470231-462EA 1929.55 CAD
470231-462 470231-464
Boreal C-SCOPE Compound Microscopes
Microscopes
A robust microscope that provides crisp and clear visual and digital results!
  • Infinity optics
  • Plan lead-free objectives, 4, 10, 40 and 100X oil immersion
  • Rackless mechanical stage
  • Custom camera packages available for Trinocular model
The new Boreal C-SCOPE models are setting a new standard in microscope performance through improvements both in optical and mechanical features. Designed to be used in educational life science, medical and a variety of biological applications, these new microscopes are built around Boreal's complete understanding of the daily demands placed on a standard educational microscope.
Student proof features together with Infinity Plan optics provide fully corrected, repeatable, quality images.
Ordering information: The camera packages for microscope 470231-464 are sold separately and include everything needed to connect to the microscope.