AlphaTec® 58-128 Light Duty Chemical Protection Gloves, Ansell
Supplier: Ansell Healthcare
58128070 58128080 58128090 58128100 58128110
75846-510CS 2390.77 CAD
75846-510 75846-512 75846-514 75846-516 75846-518
AlphaTec® 58-128 Light Duty Chemical Protection Gloves, Ansell
Gloves
First ergonomically certified chemical glove. Maximize worker productivity by minimizing hand fatigue.
  • Provides quantifiable ergonomic benefits to the wearer
  • Delivers high-performance musculoskeletal support
  • Conforms to the shape of the wearer’s hand enabling natural movement
  • Reduces the risk of ergonomic injury caused by prolonged or repetitive manual tasks
ERGOFORM™ helps support musculoskeletal health during repetitive tasks and improves worker performance.
Unique 'Thin Nitrile Technology' provides a thin barrier while offering superior protection from oils and other irritant fluids.
Ansell Grip Technology™ allows for less force and optimal control and confidence when gripping oily or wet objects, reducing hand fatigue and enhancing worker comfort.
The ergonomic design combines a flexible open weave knit liner and stretchy nitrile formulation coating with a shape that conforms to the wearer's hand. This enables natural movement, allowing for better dexterity, tactility, and comfort while reducing hand fatigue.
Certifications: Meets ANSI Abrasion Level 3.