VWR® Upright Freezer Rack for Microplates
Supplier: VWR International
76027-630 76027-632 76052-204
76027-630EA 251.45 CAD
76027-630 76027-632 76052-204
VWR® Upright Freezer Rack for Microplates
Racks Cryogenic Box Racks
Organize Freezer Samples
Racks are designed to store 96-well, SBS format plates. They are made from corrosion-resistant stainless steel.