VWR® Upright Freezer Racks for 2" Boxes
Supplier: VWR International
76027-604 76027-796 76027-798 76027-800 76027-810 76027-812 76027-814 76051-406 76051-408 76051-418 76051-422 76051-424 76530-432
76027-604EA 232.27 CAD
76027-604 76027-796 76027-798 76027-800 76027-810 76027-812 76027-814 76051-406 76051-408 76051-418 76051-422 76051-424 76530-432
VWR® Upright Freezer Racks for 2" Boxes
Racks Cryogenic Box Racks
Organize freezer samples.
  • Organize and find samples quickly
  • Space-saving design for specific freezer requirements
  • Security locking devices available
Racks are designed for use with standard 2" cardboard, plastic, or metal boxes. Racks are made from corrosion-resistant stainless steel. Security lock devices are also available.