Vertical Laminar Flow Hood, DFMZ
Supplier: DFMZ
LHV-4B LHV-5B LHV-3B LHV-5B LHV-3B LHV-4B LHV-6B LHV-6B
CA97015-418EA 24519.22 CAD
CA97015-418 CA97015-416 97015-418 CA97015-414 97015-416 97015-414 CA97015-420 97015-420
Vertical Laminar Flow Hood, DFMZ
Fume Hoods and Enclosures Laminar Flow Cabinets
LHV Laminar flow hoods are constructed from corrosion-resistant white polypropylene which is ideal for processes involving corrosive chemicals and are resilient to frequent sanitizing
  • Minimum ISO 5 (Class 100) conditions in the work zone
  • High Efficiency Particulate Air (HEPA) filter rated 99.99% efficient at 0.3 microns
  • Minihelic gauge to monitor loading on HEPA filter
  • Fluorescent task lighting
  • Disposable, quick-change pre-filters, rated MERV 7
  • Variable speed control
  • Side windows for additional viewing angles
  • Separate On/Off switches for lights and fans
It features maximum work zone depth while still fitting through most standard doorways
Ordering information: Optional accessories like Base frame (sitting or standing height), Base frame (electric hydraulic adjustable height), Vertically sliding face shield, fully counter-balanced, Stainless steel IV Bar, UV light fixture, Stainless steel lining on interior, Duplex electrical outlets, Service valves, Energy-efficient motor package andMetal-free constructions are available on requests.