Zenix™-C SEC Columns, Sepax technologies
Supplier: Sepax Technologies
233150P-4605 233150-4625 233150-4605 233100P-4605 233300-4615 233300-4625 233300-7815 233300-7820 233300-7825 233100-4625 233100-4630 233100-4605 233100-7825 233100-7830 233100-7805 233080-4615
103061-260EA 2040 CAD
103061-260 103061-264 103061-268 103061-472 103061-492 103061-494 103061-500 103061-502 103061-504 103061-532 103061-534 103061-536 103061-540 103061-542 103061-544 103061-590
Zenix™-C SEC Columns, Sepax technologies
Chromatography Columns
Zenix™-C SEC phase, developed based on innovative surface coating technology, comprises of uniform, hydrophilic, and neutral nanometer thick film chemically bonded to high purity and mechanically stabilized silica.
  • Highest separation efficiency and resolution
  • High capacity and high lot-to-lot reproducibility
  • Aqueous and organic mobile phase compatibility
  • High protein recovery with intact biological activity
  • Negligible non-specific interactions
Zenix™-C uses lay-down monolayer on porous silica offers ideal phase chemistry for hydrophobic samples like insulin, membrane protein monoclonal antibodies derivatized with polymer branches (polypeptide or PEG) separation.
Packaging: * Column packed with PEEK tubing.