YMC-Triart C8 Hybrid Silica HPLC Columns, C8, 5 µm, YMC America
Supplier: YMC America,Inc
TO12S05-0520WX TO12S05-0530WX TO12S05-1020WX TO12S05-1030WX TO12S05-L530WX TO12S05-0310WTG TO12S05-1510WT TO12S05-1520WX TO12S05-1530WX TO12S05-2510WT TO12S05-2520WX TO12S05-2530WX TO12S05-0510WT TO12S05-0553DX TO12S05-1010WT TO12S05-1053DX
89217-098EA 5235.84 CAD
89217-098 89217-100 89217-108 89217-110 89217-128 10842-256 10842-262 10842-264 10842-266 10842-274 10842-276 10842-278 76013-936 76013-938 76013-940 76013-942
YMC-Triart C8 Hybrid Silica HPLC Columns, C8, 5 µm, YMC America
Chromatography Columns
YMC-Triart C8, 5 µm semi-preparative and preparative columns are packed with a novel, organic/inorganic hybrid silica particles derivatized with C8.
  • Suitable for HPLC
  • Same selectivity as other Triart C8 particle sizes
  • Chemically and mechanically stable
  • Base-washable (clean in place)
  • High loading capacity
The core material for all of the YMC-Triart products, the particle, is inspired by YMC’s flow chemistry technology. Inherent in YMC’s process is a high level of temperature and particle formation control. As a result, Triart particles are consistent,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable