Gold Coated Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78750-01 78750-06 78750-10 78750-11 78750-12 78750-02 78750-05 78750-04 78750-07 78750-09 78750-03 78750-08
103301-810EA 87.28 CAD
103301-810 103301-818 103302-136 103302-138 103302-144 103302-446 103302-448 103302-532 103302-542 103302-610 103302-644 103302-676
Gold Coated Tweezers, Electron Microscopy Sciences
Forceps
These gold coated tweezers are finely crafted, high precision tweezers made from anti-acidic, non-magnetic stainless steel, plated with 2 microns of pure 24-carat gold
  • Resists chemical corrosion and oxidation
  • Excellent conductor of electricity
These tweezers are autoclavable and conducive for use when working with microelectronics, TEM and immunogold staining, electro-chemistry and nanotechnology.