Certified Microcentrifuge Tubes in Self-Standing Bags, Globe Scientific
Supplier: Globe Scientific
111564 111554 111554G 111554N 111554B 111554R 111554Y 111554V 111554A 111564G 111564N 111564B 111564R 111564Y 111564V 111564A 111574 111574G 111574N 111574B 111574R 111574Y 111574V 111574A
75845-358BG 40.51 CAD
75845-358 76004-078 76004-080 76004-082 76004-084 76004-086 76004-088 76004-090 76004-092 76004-094 76004-096 76004-098 76004-100 76004-102 76004-104 76004-106 76004-108 76004-110 76004-112 76004-114 76004-116 76004-118 76004-120 76004-122
Certified Microcentrifuge Tubes in Self-Standing Bags, Globe Scientific
Tubes Microcentrifuge Tubes
The polypropylene (PP) tubes are resistant to most chemical reagents and can be autoclaved at 121 °C.
  • Now available in 8 attractive colors: amber, blue, green, natural, orange, red, violet, and yellow  
  • Efficient, automated production reduces cost and assures high quality
  • Tubes are made from USP Class VI medical grade polypropylene with excellent clarity
  • Supplied in resealable, tamper-evident, self standing bags
  • Tubes are graduated with frosted marking areas and piercable caps
  • Manufactured in a class 100000 cleanroom and certified to be free from RNase, DNase, DNA, and Pyrogens
Each tube is molded with precise graduation marks for accurate measurements. These microcentrifuge tubes are offered in resealable, tamper evident self-standing bags. 