Capto™ DEAE Ion Exchange Chromatography Media, Cytiva
Supplier: Cytiva
17544301
17544310
CA97067-892EA
771.85
CAD
CA97067-892
CA97067-894
Capto™ DEAE Ion Exchange Chromatography Media, Cytiva
Protein Purification
Protein Purification Resins
Capto™ DEAE is a strong anion exchange bioprocessing medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective capture and intermediate protein purification when high volume throughput is essential.
- High dynamic binding capacity at high flow raises productivity
- High volume throughput cuts process times
It withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime.