Capto™ Core 700 Multimodal Chromatography Media

Supplier: Cytiva

17548101 17548102 17548101 17548102
CA89230-828EA 644.82 CAD
CA89230-828 CA89230-830 89230-828 89230-830
Capto™ Core 700 Multimodal Chromatography Media
Protein Purification Protein Purification Resins

Capto™ core 700 is designed for intermediate purification and polishing of viruses and other large biomolecules (molecular weight >700000).


  • Novel core bead technology allows efficient capture of contaminants while target molecules are collected in the flowthrough
  • Significantly improved productivity and higher flow rates compared with gel filtration methods
  • Straightforward optimization due to flowthrough chromatography and robust performance
  • Convenient small-scale purification, process development, and scale-up using prepacked HiTrap and HiScreen columns


The novel core bead technology and multimodal, octylamine ligand give dual functionality, size exclusion, and binding chromatography in one chromatography medium (resin). Available in various pack sizes for packing columns and in convenient, prepacked formats.

Order Now

SPECIFICATIONS

Ligand Octylamine Octylamine
Environmentally Preferable
Matrix Highly cross-linked agarose Highly cross-linked agarose
Flow rate <500 cm/h <500 cm/h
pH range 3 - 13 3 - 13
Total ionic capacity 40 - 85 µmol/ml (Cl-) 40 - 85 µmol/ml (Cl-)
Binding capacity ~ 13 mg ovalbumin ~ 13 mg ovalbumin
Storage temperature 4 - 30 °C 4 - 30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR