PTFE Scoop, Bürkle
Supplier: BURKLE INC
5389-1000 5389-0001 5389-0010 5389-0100 5389-0250 5389-0500
76049-262EA 922.2 CAD
76049-262 76049-264 76049-272 76049-274 76049-276 76049-278
PTFE Scoop, Bürkle
Scoops
All parts coming into contact with the sample medium are made of pure white PTFE
The rod is stabilized with a steel core that is completely encapsulated in PTFE. The 60 cm (23.62") long rod can be extended to 120 cm (47.24") by adding the extension component. The 10 mL (0.34 oz.) PTFE scoop has a strong rod with a length of 155 mm (6.10").
Completely inert against all substances (with the exception of elementary fluoride).