Burkholderia cepacia Selective Supplement, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
FD232-5VL FD232-5X5VL
CA71005-926PK 625.51 CAD
CA71005-926 CA71005-928
Burkholderia cepacia Selective Supplement, HiMedia
Media for Microbiology Additives and Supplements for Microbiology
An antibiotic supplement recommended for the selective isolation of Burkholderia cepacia.