Sodium dodecyl sulfate (SDS) solution 20%, sterile

Supplier: TEKNOVA CA

S0294 S0295 S0293
CA100218-170EA 439.27 CAD
CA100218-170 CA100219-182 CA100219-826
Sodium dodecyl sulfate (SDS) solution 20%, sterile
Sodium dodecyl sulphate (SDS)

SDS is an anionic detergent, commonly used to solubilize proteins, lipids and lyse bacterial cells for DNA extraction and plasmid purification.

Formula: CH₃(CH₂)₁₁OSO₃Na
MDL Number: MFCD00036175
CAS Number: 151-21-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR