Rotors JS-4.750, JS-4.750µ and JA-10.100 for Avanti™ J-15 Series Benchtop Centrifuges, Beckman Coulter
Supplier: Beckman Coulter
B77584 B77580 B83980
CA76121-328EA 5094.84 CAD
CA76121-328 CA76121-330 CA76121-332
Rotors JS-4.750, JS-4.750µ and JA-10.100 for Avanti™ J-15 Series Benchtop Centrifuges, Beckman Coulter
Centrifuges Rotors
The Avanti™ J-15 series offers three rotors, the JS-4.750, JS-4.750µ and the JA-10.100.
  • Optional BioCertified Aerosolve canisters
  • Wide list of modular disk adapters
  • RCF needed for various applications
The Avanti™ J-15 Series of benchtop centrifuges offers three rotors: JS-4.750, JS-4.750µ and JS-10.100.
The JS-4.750 is a four-place swinging-bucket rotor. The rotor buckets carry adapters that allow centrifugation of a wide range of tube and bottle sizes (from 1.5 mL reaction vials to 750 mL bottles), single- to quad-pack blood bags, and multi-wellMore Product Information