SW® PowerChem® PC-95GR Green 4.7 mil Neoprene Exam Gloves
Supplier: SW Safety Solutions
K001404 K001405 K001402 K001403 K001406
76051-156BX 51 CAD
76051-156 76051-188 76051-220 76051-222 76051-224 77463-352
SW® PowerChem® PC-95GR Green 4.7 mil Neoprene Exam Gloves
Gloves
When the situation calls for high performance, these neoprene gloves provide superior resistance to chemicals, punctures, and abrasions with a secure grip for handling small parts and slippery objects.
  • Pure neoprene prevents latex allergic reactions
  • Textured surface delivers excellent grip
  • Ergonomic design for comfort
  • Optimal thickness for durability and tactile sensitivity
  • Powder-free to prevent contamination
  • Tear-resistant beaded cuff eases donning
  • Tested for use with chemotherapy drugs
  • SHA Dermatologically Approved
The SW difference: 100% air inspected, 75% less residue, consistent tip to cuff.
Born of SW® manufacturing capabilities and advancements in material science, PowerChem™ offers chemical protection for highly specific applications. Using the most advanced materials and SW®'s proven manufacturing techniques, PowerChem™ provides the optimum blend of tactility and protection in a 5 mil glove. Ergonomically designed for comfort, this 1.5 AQL glove features a fully textured surface for secure grip in a variety of wet and dry environments and is approved for use with chemotherapy gloves.
Tested for use with chemotherapy drugs, PowerChem™ is also Dermatologically Approved by the Skin Health Alliance and has met stringent standards that ensure skin safety.
Biodegradable in landfills: 92.6% biodegradable in 2.5 years. ASTM D5526-12 test results do not predict future biodegradation. EcoTek® provides all the benefits of biodegradability without sacrificing performance, safety, or durability.
Certifications: WBENC Women Owned; NCMSDC Minority Business; Small Business Enterprise; SHA Dermatologically Approved; Meets stringent standards that ensure skin safety; EAC TP TC 019/2011; CE 0493; CE EN 455.
Ordering information: Glove sizes are as follows; XS (5.5 to 6), S (6.5 to 7), M (7.5 to 8), L (8.5 to 9), XL (9.5 to 10), XXL (10.5 to 11).