VWR® Color Micro (Flea) Spinbar Magnetic Stirring Bars
Supplier: Bel-Art Products, a Part of SP
47751-494 47751-496 47751-498 47751-500 47751-502 47751-504 47751-508 47751-510 37121-6022 47751-514 37121-6024 47751-506 76001-842 76001-844 76001-846 76001-848 76001-850 76001-852 76001-854 76001-856 76001-858 76001-860 76001-862 76001-864 76001-866 76001-898 76001-900
47751-494EA 11.38 CAD
47751-494 47751-496 47751-498 47751-500 47751-502 47751-504 47751-508 47751-510 47751-512 47751-514 47751-516 47751-506 76001-842 76001-844 76001-846 76001-848 76001-850 76001-852 76001-854 76001-856 76001-858 76001-860 76001-862 76001-864 76001-866 76001-898 76001-900
VWR® Color Micro (Flea) Spinbar Magnetic Stirring Bars
Stirrers Magnetic Stirring Bars
Micro (Flea) magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes and gradient makers.
  • Available in a variety of colors and sizes
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated.