SW® PowerForm® PF-95BL Blue 6.1 mil Nitrile Exam Dry Core® Sweat Absorbent Gloves
Supplier: SW Safety Solutions
N106552 N106553 N106554 N106555 N106556
76050-998BX 36.53 CAD
76050-998 76051-000 76051-002 76051-004 76051-006 76051-008
SW® PowerForm® PF-95BL Blue 6.1 mil Nitrile Exam Dry Core® Sweat Absorbent Gloves
Gloves
Keep hands cooler and more comfortable with premium quality nitrile gloves, featuring Unique Dry Core™ moisture management technology.
  • Dry Core® moisture management technology pulls sweat away from the skin leaving the wearer dryer
  • Fully textured surface for superior wet and oily grip
  • Silicone-free for critical environments
  • 1.5 AQL exceeds laboratory standards
  • ANSI level 3 abrasion
  • EcoTek® sustainable technology
EcoTek® gloves tested to ASTM D5526-12 standard test method for determining anaerobic biodegradation of plastic materials under accelerated landfill conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated. For additional information on the test reports, contact cs@swssglobal.com.
Certifications: WBENC Women Owned; NCMSDC Minority Business; Small Business Enterprise; SHA Dermatologically Approved; Meets stringent standards that ensure skin safety; EAC TP TC 019/2011; CE 0493; CE EN 455; ANSI Level 3 Abrasion.
Ordering information: Glove sizes are as follows; S (6.5 to 7), M (7.5 to 8), L (8.5 to 9), XL (9.5 to 10), XXL (10.5 to 11).