VWR® Reusable Volumetric Class B Color-Coded Pipettes
Supplier: VWR International
122.422.08 122.422.01 122.422.02 122.422.02A 122.422.03 122.422.04 122.422.05 122.422.06 122.422.07
75998-012CS 355.11 CAD
75998-012 75998-048 75998-050 75998-052 75998-054 75998-056 75998-058 75998-060 75998-062
VWR® Reusable Volumetric Class B Color-Coded Pipettes
Pipettes Graduated Pipettes
VWR® volumetric pipettes are made from Borosilicate glass, ASTM 438, Type 1, Class B standard.
  • Precise temperature
  • Accurate measurement
  • Measure delivery of liquid volumes
These graduated pipettes are calibrated to deliver a precise volume at a particular temperature.