VWR® Reusable Mohr-Style, Color-Coded, TD Pipettes, Class B
Supplier: VWR International
75998-032 75998-034 75998-036 75998-038 75998-040 75998-042 75998-044 75998-046
75998-032CS 275.64 CAD
75998-032 75998-034 75998-036 75998-038 75998-040 75998-042 75998-044 75998-046
VWR® Reusable Mohr-Style, Color-Coded, TD Pipettes, Class B
Pipettes Measuring (Mohr) Pipettes
VWR® Mohr pipettes are made from Borosilicate glass, ASTM 438, Type 1, Class B standard.
  • Designed as straight glass tubes
  • With one tapering end
  • They are calibrated into small divisions
  • Various amounts of liquid can be measured within the same device
  • Mohr pipettes are calibrated to deliver a precise volume at a precise temperature
  • They are used for accurate measurement and delivery of liquid volumes
The VWR® Mohr Pipettes Calibration and toleration standards for these pipettes are defined as per standard specifications and practices: ASTM E1044, ASTM E1293, ASTM E542, and ASTM E694.