YMC-Triart C8 Hybrid Silica HPLC Columns, C8, 5 µm, YMC America
Supplier: YMC America,Inc
TO12S05-0202WT TO12S05-1504WT TO12S05-1506WT TO12S05-2503WT TO12S05-2504WT TO12S05-2506WT TO12S05-3006WT TO12S05-H546WT TO12S05-R503WT TO12S05-R504WT TO12S05-1546PTP TO12S05-05Q1PTP TO12S05-10Q1PTP TO12S05-15Q1PTP TO12S05-0546PTP TO12S05-1046PTP TO12S05-0504WT
10842-248EA 1371.76 CAD
10842-248 10842-258 10842-260 10842-268 10842-270 10842-272 10842-280 10842-282 10842-284 10842-286 10842-288 75995-060 75995-160 75995-162 75995-164 75995-166 75995-168 76013-934
YMC-Triart C8 Hybrid Silica HPLC Columns, C8, 5 µm, YMC America
Chromatography Columns
YMC-Triart C8, 5µm is a novel organic/inorganic hybrid silica particle derivatized with C8 (L7 USP designation).
  • Suitable for HPLC
  • Chemical and mechanical durability of the particles contributes to long column lifetime
  • Proprietary end capping makes for excellent peak shapes of all types of compounds
  • Rigorous manufacturing controls result in legendary YMC column-to-column and lot-to-lot reproducibility
  • More retentive than other hybrid materials, potentially simplifying the separation of 'difficult' peaks
  • YMC-Triart C8 is available in packed columns of
More Product Information