YMC-Triart C8 Hybrid Silica UHPLC Columns, C8, 1.9 µm, YMC America
Supplier: YMC America,Inc
TO12SP9-0302PT TO12SP9-0502PT TO12SP9-0503PT TO12SP9-1002PT TO12SP9-1003PT TO12SP9-L502PT TO12SP9-L503PT TO12SP9-0202PT TO12SP9-0303PT TO12SP9-0546PT TO12SP9-1502PT TO12SP9-1503PT TO12SP9-05Q1PTP TO12SP9-10Q1PTP TO12SP9-15Q1PTP TO12SP9-03Q1PT TO12SP9-05Q1PT TO12SP9-15Q1PT TO12SP9-L5Q1PT
89217-132EA 2459.4 CAD
89217-132 89217-134 89217-136 89217-138 89217-140 89217-142 89217-144 10842-290 10842-292 10842-294 10842-296 10842-298 75995-142 75995-144 75995-146 76013-944 76013-946 76013-948 76013-950
YMC-Triart C8 Hybrid Silica UHPLC Columns, C8, 1.9 µm, YMC America
Chromatography Columns
YMC-Triart C8 is a novel organic/inorganic hybrid silica particle derivatized with C8 (L7 USP designation).
  • Suitable for UHPLC
  • Chemical and mechanical durability of the particles contributes to long column lifetime
  • Proprietary end capping makes for excellent peak shapes of all types of compounds
  • Rigorous manufacturing controls result in legendary YMC column-to-column and lot-to-lot reproducibility
  • More retentive than other hybrid materials, potentially simplifying the separation of 'difficult' peaks
  • YMC-Triart C8 is available in packed columns of various
More Product Information