YMC-Triart C18 S Prep Hybrid Silica HPLC Columns, C18 S Prep, SemiPrep/Preparative, YMC America
Supplier: YMC America,Inc
TAS12S11-0520WX TAS12S11-0530WX TAS12S11-1010WT TAS12S11-1020WX TAS12S11-1030WX TAS12S11-1053DX TAS12S11-1510WT TAS12S11-1520WX TAS12S11-1530WX TAS12S11-1553DX TAS12S11-2520WX TAS12S11-L530WX TAS12S16-0520WX TAS12S21-0520WX TAS12S11-0310WT TAS12S11-0510WT TAS12S11-0553DX TAS12S11-2510WT TAS12S11-2530WX TAS12S11-2553DX
10842-076EA 5236.21 CAD
10842-076 10842-078 10842-106 10842-108 10842-110 10842-112 10842-114 10842-116 10842-118 10842-120 10842-122 10842-128 10842-130 10842-132 76013-890 76013-892 76013-902 76013-910 76013-912 76013-914
YMC-Triart C18 S Prep Hybrid Silica HPLC Columns, C18 S Prep, SemiPrep/Preparative, YMC America
Chromatography Columns
YMC-Triart C18, 10, 15 and 20 µm semi-preparative and preparative columns are packed with a novel, organic/inorganic hybrid silica particles derivatized with C18.
  • Same selectivity as analytical Triart C18 particle sizes
  • Chemically and mechanically stable
  • Base-washable (clean in place)
  • Particle sizes of 10, 15, and 20 µm
  • Ability to operate in 100% aqueous environments
  • High loading capacity
The core material for all of the YMC-Triart products, the particle, is inspired by YMC’s flow chemistry technology. Inherent in YMC’s process is a high level ofMore Product Information