YMC-Triart C18 S Prep Hybrid Silica HPLC Columns, C18 S Prep, 10 µm, Analytical, YMC America
Supplier: YMC America,Inc
TAS12S11-2546WT TAS12S11-3004WT TAS12S11-0308WTG TAS12S11-1504WT TAS12S11-1546WT TAS12S11-3008WT
10842-124EA 2773.6 CAD
10842-124 10842-126 76013-888 76013-906 76013-908 76013-916
YMC-Triart C18 S Prep Hybrid Silica HPLC Columns, C18 S Prep, 10 µm, Analytical, YMC America
Chromatography Columns
YMC-Triart C18 S Prep, 10 µm columns are packed with a novel, organic/inorganic hybrid silica particles derivatized with C18.
  • Ideal for HPLC
  • Same selectivity as other Triart C18 particle sizes
  • Chemically and mechanically stable
  • Base-washable (clean in place)
  • Ability to operate in 100% aqueous environments
The core material for all of the YMC-Triart products, the particle, is inspired by YMC’s flow chemistry technology. Inherent in YMC’s process is a high level of temperature and particle formation control. As a result, Triart particles are consistent,More Product Information