Helix Elite Synthetic Standard, Microbiologics
Supplier: MICROBIOLOGICS, INC.
HE0012S HE0013S
10753-270EA 780.55 CAD
10753-270 10753-272 10753-274 10753-276 10753-278 10753-280 10753-282 10753-284 10753-286 10753-288 10753-290 10753-292 10753-294 10753-296 10753-298
Helix Elite Synthetic Standard, Microbiologics
Standards Biological Standards
Comopatible with a variety of instruments, kits, and applications.
Synthetic nucleic acid RNA and DNA sequences for use as amplification and detection controls in clinical assays
Each standard contains consensus sequences of diagnostic regions from the target’s genome, representing the known genetic diversity of the microorganism; ready-to-use with no extraction required
Compatible with a variety of instruments, kits and applications
Each Synthetic Helix Elite Molecular Standard contains consensus sequences of diagnostic regions from the target’s genome, representing the known genetic diversity of the microorganism; ready-to-use with no extraction required. This standard is compatible with a variety of instruments, kits and applications.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the product page. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.