Whatman™ pH Indicator Papers, Reels, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10362030 10362030
CA11013-424PK 54.98 CAD
CA11013-424 10034-828
Whatman™ pH Indicator Papers, Reels, Whatman products (Cytiva)
Test Papers pH Indicator Papers and Test Strips
Universal indicator papers which have been impregnated with a mixture of several indicators.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use
On contact with the sample solution they assume a particular color. A check against the color comparision table supplied allows the pH to be determined.