YMC-Triart C18 ExRS Hybrid Silica HPLC Columns, C18 ExRS, 5 µm, YMC America
Supplier: YMC America,Inc
TAR08S05-02Q1PTH TAR08S05-0503PTH TAR08S05-0546PTH TAR08S05-05Q1PTH TAR08S05-1003PTH TAR08S05-1046PTH TAR08S05-10Q1PTH TAR08S05-1503PTH TAR08S05-1546PTH TAR08S05-15Q1PTH TAR08S05-2546PTH TAR08S05-H346PTH TAR08S05-H3Q1PTH TAR08S05-L503PTH TAR08S05-L546PTH TAR08S05-L5Q1PTH TAR08S05-05Q1PTP TAR08S05-10Q1PTP TAR08S05-15Q1PTP TAR08S05-0546PTP TAR08S05-1046PTP TAR08S05-1546PTP
10498-574EA 1300.5 CAD
10498-574 10498-576 10498-578 10498-580 10498-582 10498-584 10498-586 10498-588 10498-590 10498-592 10498-594 10498-596 10498-598 10498-600 10498-602 10498-604 75995-048 75995-050 75995-052 75995-054 75995-056 75995-140
YMC-Triart C18 ExRS Hybrid Silica HPLC Columns, C18 ExRS, 5 µm, YMC America
Chromatography Columns
YMC-Triart C18 ExRS, 5µm columns are based on technology featuring an organic/inorganic hybrid silica particle that utilizes polymeric bonding end capping.
  • C18 phase bonded to organic/inorganic hybrid silica gel
  • Excellent selectivity of isomers and structural analogs
  • Superior chemical durability
  • Identical selectivity over particle sizes
  • Ideal for HPLC
The core material for all of the YMC-Triart products, the particle, is inspired by YMC’s flow chemistry technology. Inherent in YMC’s process is a high level of temperature and particle formation control. As a result, Triart particles are consistent, reproducible, and scalable across many particle sizes. The hybrid Triart particles produce a material that is highly resilient and robust when used at extreme pH and elevated temperature. These particles are very stable under mechanical stress such as high pressure. YMC-Triart C18 ExRS can be used at temperatures up to 70 °C at pH 1 to 7, and 50 ⁰C from pH 7 to 12.
In the case of Triart ExRS, hydrophobicity is high due to the high carbon loading (25%). Triart ExRS suitable for use with separation of isomers and structural analogs.