YMC-Triart C18 ExRS Hybrid Silica HPLC Columns, C18 ExRS, 3 µm, YMC America
Supplier: YMC America,Inc
TAR08S03-02Q1PTH TAR08S03-0503PTH TAR08S03-0546PTH TAR08S03-05Q1PTH TAR08S03-1003PTH TAR08S03-1046PTH TAR08S03-10Q1PTH TAR08S03-1503PTH TAR08S03-1546PTH TAR08S03-15Q1PTH TAR08S03-2546PTH TAR08S03-H346PTH TAR08S03-H3Q1PTH TAR08S03-L503PTH TAR08S03-L546PTH TAR08S03-L5Q1PTH TAR08S03-05Q1PTP TAR08S03-10Q1PTP TAR08S03-15Q1PTP TAR08S03-0546PTP TAR08S03-1046PTP TAR08S03-1546PTP TAR08S03-2503PTH
10498-390EA 1323.87 CAD
10498-390 10498-392 10498-394 10498-396 10498-398 10498-400 10498-402 10498-404 10498-406 10498-560 10498-562 10498-564 10498-566 10498-568 10498-570 10498-572 75995-036 75995-038 75995-040 75995-042 75995-044 75995-046 76013-880
YMC-Triart C18 ExRS Hybrid Silica HPLC Columns, C18 ExRS, 3 µm, YMC America
Chromatography Columns
YMC-Triart C18 ExRS, 3 µm columns are based on technology featuring an organic/inorganic hybrid silica particle that utilizes polymeric bonding end capping.
  • C18 phase bonded to organic/inorganic hybrid silica gel
  • Excellent selectivity of isomers and structural analogs
  • Superior chemical durability
  • Identical selectivity over particle sizes
  • Ideal for HPLC
The core material for all of the YMC-Triart products, the particle, is inspired by YMC's flow chemistry technology. Inherent in YMC’s process is a high level of temperature and particle formation control. As a result, Triart particles are consistent, reproducible, and scalable across many particle sizes. The hybrid Triart particles produce a material that is highly resilient and robust when used at extreme pH and elevated temperature. These particles are very stable under mechanical stress such as high pressure. YMC-Triart C18 ExRS can be used at temperatures up to 70 °C at pH 1 to 7, and 50 °C from pH 7 to 12.
In the case of Triart ExRS, hydrophobicity is high due to the high carbon loading (25%). Triart ExRS suitable for use with separation of isomers and structural analogs.