YMC-Triart C18 ExRS Hybrid Silica UHPLC Columns, C18 ExRS, 1.9 µm, YMC America
Supplier: YMC America,Inc
TAR08SP9-0503PT TAR08SP9-1502PT TAR08SP9-L503PT TAR08SP9-0302PT TAR08SP9-0502PT TAR08SP9-L502PT TAR08SP9-0202PT TAR08SP9-1002PT TAR08SP9-0303PT TAR08SP9-05Q1PTP TAR08SP9-10Q1PTP TAR08SP9-15Q1PTP TAR08SP9-1003PT TAR08SP9-10Q1PT TAR08SP9-1503PT
10661-972EA 2354.6 CAD
10661-972 10661-978 10661-980 10661-982 10661-984 10661-986 10662-116 10662-118 10662-120 75995-030 75995-032 75995-034 76013-882 76013-884 76013-886
YMC-Triart C18 ExRS Hybrid Silica UHPLC Columns, C18 ExRS, 1.9 µm, YMC America
Chromatography Columns
YMC-Triart C18 ExRS, 1.9 µm columns are based on technology featuring an organic/inorganic hybrid silica particle that utilizes polymeric bonding end capping.
  • C18 phase bonded to organic/inorganic hybrid silica gel
  • Excellent selectivity of isomers and structural analogs
  • Superior chemical durability
  • Identical selectivity over particle sizes
  • Ideal for UHPLC
The core material for all of the YMC-Triart products, the particle, is inspired by YMC's flow chemistry technology. Inherent in YMC’s process is a high level of temperature and particle formationMore Product Information