YMC-Triart SIL Hybrid Silica HPLC Columns, Triart SIL, 3 µm, YMC America
Supplier: YMC America,Inc
TS12S03-1046WT TS12S03-1503WT TS12S03-2546WT
76027-732EA 1723.56 CAD
76027-732 76027-734 76027-740
YMC-Triart SIL Hybrid Silica HPLC Columns, Triart SIL, 3 µm, YMC America
Chromatography Columns
YMC-Triart Silica columns are intended for use in normal phase and HILIC chromatography modes.
  • Bare organic/inorganic hybrid silica particle
  • Identical selectivity over particle sizes
  • Ideal for HPLC
  • May be used in normal phase or HILIC chromatography modes
YMC-Triart Silica is available in columns packed with particle sizes of 1.9, 3, 5, 10, 15, and 20 µm, intended for use on both SFC and HPLC systems.
Columns are available in many sizes. Please inquire if you do not see the column geometry you desire.