YMC-Triart Phenyl Hybrid Silica HPLC Columns, Triart Phenyl, 5 µm, Analytical, YMC America
Supplier: YMC America,Inc
TPH12S05-1001WT TPH12S05-3004WT TPH12S05-05Q1PTP TPH12S05-10Q1PTP TPH12S05-15Q1PTP TPH12S05-0546PTP TPH12S05-1046PTP TPH12S05-1546PTP TPH12S05-2503WT TPH12S05-H346WT
10842-362EA 1592.89 CAD
10842-362 10842-366 75995-080 75995-082 75995-170 75995-172 75995-174 75995-176 76027-716 76027-724
YMC-Triart Phenyl Hybrid Silica HPLC Columns, Triart Phenyl, 5 µm, Analytical, YMC America
Chromatography Columns
Phenyl columns often provide unique separations compared to aliphatic phases (ex).
  • Phenyl phase bonded to organic/inorganic hybrid silica particle
  • Different selectivity from C18 or C8 with polar interaction and π-π interaction playing a significant role in the separation
  • Superior chemical durability
  • Identical selectivity over particle sizes
  • Ideal for HPLC
  • Effective for separation of compounds with long conjugated unsaturated systems
YMC-Triart Phenyl can be used at temperatures up to 50 °C at pH ranging from 1 to 10. This allows the chromatographer to choose a wide range of pH and temperature conditions as additional tools to produce separations with adequate retention and separation.
C18, C8, and C4) as interactions between the analyte of interest and the phenyl stationary phase often result in unique selectivity for aromatic molecules.