YMC-Triart Phenyl Hybrid Silica HPLC Columns, Triart Phenyl, 3 µm, YMC America
Supplier: YMC America,Inc
TPH12S03-1001WT TPH12S03-05Q1PTP TPH12S03-10Q1PTP TPH12S03-15Q1PTP TPH12S03-0546PTP TPH12S03-1046PTP TPH12S03-1546PTP TPH12S03-05Q1WT
10842-352EA 1844 CAD
10842-352 75995-068 75995-070 75995-072 75995-074 75995-076 75995-078 76027-702
YMC-Triart Phenyl Hybrid Silica HPLC Columns, Triart Phenyl, 3 µm, YMC America
Chromatography Columns
Phenyl columns often provide unique separations compared to aliphatic phases.
  • Phenyl phase bonded to organic/inorganic hybrid silica particle
  • Different selectivity from C18 or C8 with polar interaction and π-π interaction playing a significant role in the separation
  • Superior chemical durability
  • Identical selectivity over particle sizes
  • Ideal for HPLC
  • Effective for separation of compounds with long conjugated unsaturated systems
YMC-Triart Phenyl can be used at temperatures up to 50 °C at pH ranging from 1 to 10. This allows the chromatographer to choose a wide range of pH and temperature conditions as additional tools to produce separations with adequate retention and separation.
C18, C8, and C4) as interactions between the analyte of interest and the phenyl stationary phase often result in unique selectivity for aromatic molecules.