YMC-Triart SIL Hybrid Silica HPLC Columns, Triart SIL, 5 µm, Analytical, YMC America
Supplier: YMC America,Inc
TS12S05-1546WT TS12S05-2546WT TS12S05-1046WT
76027-748EA 1932.62 CAD
76027-748 76027-756 76027-786
YMC-Triart SIL Hybrid Silica HPLC Columns, Triart SIL, 5 µm, Analytical, YMC America
Chromatography Columns
YMC-Triart Silica columns are intended for use in normal phase and HILIC chromatography modes.
  • Bare organic/inorganic hybrid silica particle
  • Identical selectivity over particle sizes
  • Ideal for HPLC
  • May be used in normal phase or HILIC chromatography modes
YMC-Triart Silica is available in columns packed with particle sizes of 1.9, 3, 5, 10, 15, and 20 µm, intended for use on both SFC and HPLC systems.
Columns are available in many sizes. Please inquire if you do not see the column geometry you desire.