YMC-Triart PFP Hybrid Silica HPLC Columns, PFP, 5 µm, Semi-Preparative/Prep, YMC America
Supplier: YMC America,Inc
TPF12S05-0510WT TPF12S05-0520WX TPF12S05-0530WX TPF12S05-0553DX TPF12S05-1010WT TPF12S05-1020WX TPF12S05-1030WX TPF12S05-1053DX TPF12S05-1520WX TPF12S05-1530WX TPF12S05-1553DX TPF12S05-2520WX TPF12S05-2530WX TPF12S05-2553DX TPF12S05-L530WX
76027-588EA 1985.65 CAD
76027-588 76027-590 76027-592 76027-594 76027-596 76027-668 76027-670 76027-672 76027-674 76027-676 76027-678 76027-682 76027-684 76027-686 76027-688
YMC-Triart PFP Hybrid Silica HPLC Columns, PFP, 5 µm, Semi-Preparative/Prep, YMC America
Chromatography Columns
YMC-Triart PFP is a non-endcapped stationary phase with unique selectivity.
  • Pentafluorophenyl phase bonded to organic/inorganic hybrid silica particle
  • Different selectivity from C18 or C8 with polar interaction and π-π interaction playing a significant role in the separation
  • Superior chemical durability
  • Identical selectivity over particle sizes
  • Ideal for HPLC
  • Superior planar and steric cognitive ability
YMC-Triart PFP can be used at temperatures up to 50 °C at pH 1 to 8. This allows the chromatographer to choose a wide range of pH and temperature conditions as additional tools to produce separations with adequate retention and separation.
It is sometimes useful for separation of halogen-containing compounds.