YMC-Triart PFP Hybrid Silica HPLC Columns, PFP, 5 µm, Analytical, YMC America
Supplier: YMC America,Inc
TPF12S05-05Q1PTP TPF12S05-10Q1PTP TPF12S05-15Q1PTP TPF12S05-0546PTP TPF12S05-1046PTP TPF12S05-1546PTP TPF12S05-2503WT
75995-196EA 2626.8 CAD
75995-196 75995-198 75995-200 75995-202 75995-204 75995-206 76027-680
YMC-Triart PFP Hybrid Silica HPLC Columns, PFP, 5 µm, Analytical, YMC America
Chromatography Columns
YMC-Triart PFP is a non-endcapped stationary phase with unique selectivity.
  • Pentafluorophenyl phase bonded to organic/inorganic hybrid silica particle
  • Different selectivity from C18 or C8 with polar interaction and π-π interaction playing a significant role in the separation
  • Superior chemical durability
  • Identical selectivity over particle sizes
  • Ideal for HPLC
  • Superior planar and steric cognitive ability
YMC-Triart PFP can be used at temperatures up to 50 °C at pH 1 to 8. This allows the chromatographer to choose a wide range of pH and temperature conditions as additional tools to produce separations with adequate retention and separation.
It is sometimes useful for separation of halogen-containing compounds.