YMC-Triart PFP Hybrid Silica HPLC Columns, PFP, 3 µm, YMC America
Supplier: YMC America,Inc
TPF12S03-05Q1PTP TPF12S03-10Q1PTP TPF12S03-15Q1PTP TPF12S03-0546PTP TPF12S03-1046PTP TPF12S03-1546PTP
75995-184EA 2626.78 CAD
75995-184 75995-186 75995-188 75995-190 75995-192 75995-194
YMC-Triart PFP Hybrid Silica HPLC Columns, PFP, 3 µm, YMC America
Chromatography Columns
YMC-Triart PFP is a non-endcapped stationary phase with unique selectivity.
  • Pentafluorophenyl phase bonded to organic/inorganic hybrid silica particle
  • Different selectivity from C18 or C8 with polar interaction and π-π interaction playing a significant role in the separation
  • Superior chemical durability
  • Identical selectivity over particle sizes
  • Ideal for HPLC
  • Superior planar and steric cognitive ability
YMC-Triart PFP can be used at temperatures up to 50 °C at pH 1 to 8. This allows the chromatographer to choose a wide range of pH and temperatureMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable