YMC-Triart PFP Hybrid Silica UHPLC Columns, PFP, 1.9 µm, YMC America
Supplier: YMC America,Inc
TPF12SP9-05Q1PTP TPF12SP9-10Q1PTP TPF12SP9-15Q1PTP TPF12SP9-0303PT TPF12SP9-03Q1PT TPF12SP9-05Q1PT TPF12SP9-10Q1PT TPF12SP9-15Q1PT TPF12SP9-L5Q1PT
75995-178EA 2959.07 CAD
75995-178 75995-180 75995-182 76027-690 76027-692 76027-694 76027-696 76027-698 76027-700
YMC-Triart PFP Hybrid Silica UHPLC Columns, PFP, 1.9 µm, YMC America
Chromatography Columns
YMC-Triart PFP is a non-endcapped stationary phase with unique selectivity.
  • Pentafluorophenyl phase bonded to organic/inorganic hybrid silica particle
  • Different selectivity from C18 or C8 with polar interaction and π-π interaction playing a significant role in the separation
  • Superior chemical durability
  • Identical selectivity over particle sizes
  • Ideal for UHPLC
  • Superior planar and steric cognitive ability
YMC-Triart PFP can be used at temperatures up to 50 °C at pH 1 to 8. This allows the chromatographer to choose pH and temperature as additional tools to produce baseline separations.
It is sometimes useful for separation of halogen-containing compounds.