YMC-Triart Diol-HILIC Hybrid Silica HPLC Columns, Diol-HILIC, 5 µm, Semi-Preparative/Preparative, YMC America
Supplier: YMC America,Inc
TDH12S05-0530WX TDH12S05-0553DX TDH12S05-1010WT TDH12S05-1020WX TDH12S05-2520WX TDH12S05-2530WX TDH12S05-2553DX TDH12S05-0510WT TDH12S05-0520WX TDH12S05-1030WX TDH12S05-1053DX TDH12S05-1520WX TDH12S05-1530WX TDH12S05-1553DX
76012-976EA 11122.77 CAD
76012-976 76012-978 76012-980 76012-982 76013-012 76013-014 76013-016 76013-918 76013-920 76013-922 76013-924 76013-926 76013-928 76013-930
YMC-Triart Diol-HILIC Hybrid Silica HPLC Columns, Diol-HILIC, 5 µm, Semi-Preparative/Preparative, YMC America
Chromatography Columns
Triart Diol-HILIC is a dihydroxypropane moiety bonded to an organic/inorganic hybrid based silica particle.
  • Dihydroxypropyl phase bonded to organic/inorganic hybrid silica particle
  • Ideal for the separation of polar and hydrophilic compounds that are not retained by reversed phase chromatography
  • Superior chemical durability
  • Identical selectivity over particle sizes
  • Ideal for HPLC
YMC-Triart Diol HILIC can retain extremely polar compounds while using eluents and buffers suitable for LC-MS.
The resulting product has excellent durability and usability across the wide pH range of pH 2 to 10. Triart Diol-HILIC is ideal for a separation of polar and hydrophilic compounds which are not retained by reversed phase (C18, C8, and others) chromatography.