VWR® PureTouch Cut-Resistant Glove Liners

Supplier: VWR International
76102-378 76102-380 76102-382 76102-384
76102-378BG 407.29 CAD
76102-378 76102-380 76102-382 76102-384
VWR® PureTouch Cut-Resistant Glove Liners
Gloves
Glove liners that are cut resistant and provide a comfortable barrier that enhances safety while reducing perspiration and wicking moisture during extended use.

  • Compatible underglove for all cleanroom requirements
  • 13-gauge white nylon with HPPE
  • Ambidextrous
  • Color-coded for visual size identification
  • Sturdy, launderable, and reusable

For use with latex, nitrile, PVC, and all exam gloves.

These glove liners prevent skin irritation without diminishing tactile sensitivity, and limits wear and tear.

Certifications: ANSI Cut Level 2.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR