VWR® Measuring Cylinder, Hexagonal Base, with Stopper, Class A, Serialized
Supplier: VWR International
76003-698 76003-700 76003-702 76003-704 76003-706 76003-708 76003-710 76003-712 76003-730 76003-732 76003-734 76003-736 76003-738 76003-740 76003-742 76003-744
76003-698CS 680.47 CAD
76003-698 76003-700 76003-702 76003-704 76003-706 76003-708 76003-710 76003-712 76003-730 76003-732 76003-734 76003-736 76003-738 76003-740 76003-742 76003-744
VWR® Measuring Cylinder, Hexagonal Base, with Stopper, Class A, Serialized
Cylinders Measuring Cylinders
VWR® Measuring Cylinders are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Class A serialized measuring cylinders with a hexagonal base
  • Calibrated 'to Deliver'
  • Cylinders are made to meet ASTM E 1272 norms
  • Printed double scale
VWR® Measuring Cylinders are used for the precise measurement liquid volumes. Cylinders have a large hexagonal base to increase stability. Calibration and tolerances are in compliance to standards: ASTM E1272, ASTM E542, and ASTM E694.
Certifications: Cylinders are supplied with individual work certificates.