VWR® Volumetric Flask, Clear Glass, Wide Neck, Class A, Unserialized
Supplier: VWR International
76003-892 76003-894 76003-896 76003-898 76003-900 76003-902 76003-904 76003-906 76003-908 76003-910 76003-912
76003-892CS 485.64 CAD
76003-892 76003-894 76003-896 76003-898 76003-900 76003-902 76003-904 76003-906 76003-908 76003-910 76003-912
VWR® Volumetric Flask, Clear Glass, Wide Neck, Class A, Unserialized
Flasks Volumetric Flasks
VWR® flasks are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Manufactured per ASTM E 288 Class specification
  • White diamond enamel graduation lines
  • Calibrated 'to Contain'
VWR® Volumetric Flasks are used for precise dilutions and preparations of standard solutions. These flat bottom flasks have a pear-shaped lower area with a long neck. Calibration and tolerances are in compliance to standards: ASTM E288, ASTM E542, and ASTM E792.
Certifications: Flasks are supplied with Batch certificates.