VWR® Volumetric Flask, Clear Glass, Narrow Neck, Class A, Unserialized
Supplier: VWR International
134.478.05 134.478.06 134.478.07 134.478.08 134.478.00 134.478.00B 134.478.01 134.478.02 134.478.04
76003-664CS 575.59 CAD
76003-664 76003-666 76003-668 76003-670 76003-882 76003-884 76003-886 76003-888 76003-890
VWR® Volumetric Flask, Clear Glass, Narrow Neck, Class A, Unserialized
Flasks Volumetric Flasks
VWR® flasks are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Manufactured per ASTM E 288 Class specification
  • White diamond enamel graduation lines
  • Calibrated 'to Contain'
VWR® Volumetric Flasks are used for precise dilutions and preparations of standard solutions. These flat bottom flasks have a pear-shaped lower area with a long neck. Calibration and tolerances are in compliance to standards: ASTM E288, ASTM E542, and ASTM E792.
Certifications: Flasks are supplied with Batch certificates.