VWR® Volumetric Flask, Amber, Wide Neck, Class A, Unserialized
Supplier: VWR International
131.232.03A 131.232.04A 131.232.05A 131.232.06A 131.232.07A 131.232.08A 131.232.09A 131.232.10A 131.232.11A 131.478.01A 131.478.04A 131.478.05A 131.478.06A 131.232.01A 131.232.02A 131.234.01A 131.234.02A 131.234.03A 131.234.04A 131.234.05A 131.234.06A 131.234.07A 131.234.08A 131.234.09A 131.478.07A 131.478.08A 131.478.09A 131.478.10A 131.478.11A 131.234.10A 131.234.11A
76003-616CS 508.42 CAD
76003-616 76003-618 76003-620 76003-622 76003-624 76003-626 76003-628 76003-630 76003-632 76003-770 76003-772 76003-774 76003-776 76003-786 76003-788 76003-826 76003-828 76003-830 76003-832 76003-834 76003-836 76003-838 76003-840 76003-842 76003-956 76003-958 76003-960 76003-962 76003-964 76004-062 76004-064
VWR® Volumetric Flask, Amber, Wide Neck, Class A, Unserialized
Flasks Volumetric Flasks
VWR® flasks are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Manufactured per ASTM E 288 Class specification
  • White diamond enamel graduation lines
  • Calibrated 'to Contain'
VWR® volumetric flasks are used for precise dilutions and preparations of standard solutions. These flat bottom flasks have a pear-shaped lower area with a long neck. Calibration and tolerances are in compliance to standards: ASTM E288, ASTM E542, and ASTM E792.
Certifications: Flasks are supplied with Batch certificates.