VWR® Volumetric Flask, Amber, Narrow Neck, Class A, Unserialized
Supplier: VWR International
76003-600 76003-602 76003-604 76003-606 76003-608 76003-610 76003-612 76003-614 76003-656 76003-658 76003-660 76003-662 76003-754 76003-756 76003-758 76003-760 76003-762 76003-764 76003-766 76003-768 76003-782 76003-784 76003-816 76003-818 76003-820 76003-822 76003-824 76003-954 76017-938
76003-600CS 441.5 CAD
76003-600 76003-602 76003-604 76003-606 76003-608 76003-610 76003-612 76003-614 76003-656 76003-658 76003-660 76003-662 76003-754 76003-756 76003-758 76003-760 76003-762 76003-764 76003-766 76003-768 76003-782 76003-784 76003-816 76003-818 76003-820 76003-822 76003-824 76003-954 76017-938
VWR® Volumetric Flask, Amber, Narrow Neck, Class A, Unserialized
Flasks Volumetric Flasks
VWR® flasks are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Manufactured per ASTM E 288 Class specification
  • White diamond enamel graduation lines
  • Calibrated 'to Contain'
VWR® Volumetric Flasks are used for precise dilutions and preparations of standard solutions. These flat bottom flasks have a pear-shaped lower area with a long neck. Calibration and tolerances are in compliance to standards: ASTM E288, ASTM E542, and ASTM E792.
Certifications: Flasks are supplied with Batch certificates.