VWR® Burette, Glass Stopcock, Class A, Unserialized
Supplier: VWR International
76003-576 76003-578 76003-580 76003-582
76003-576CS 170.5 CAD
76003-576 76003-578 76003-580 76003-582
VWR® Burette, Glass Stopcock, Class A, Unserialized
Burettes
VWR® Burettes are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 heat-resistant glass.
  • All burettes have a bore size of 2 mm
  • Calibrated to deliver (TD, EX) at 20 °C
  • Calibrated to meet ASTM E 287 specifications
  • All burettes are calibrated according to Class A norms
  • A batch certificate is included
VWR® unserialized Class A burettes are used to deliver solution in precisely measured, variable volumes. They are used primarily for titration, to deliver one reactant until the precise endpoint is reached. Calibration and tolerances are in compliance to standards: ASTM 3287, ASTM E542, and ASTM E694.
Certifications: Burettes are supplied with Batch certificates.