VWR® Burette, Glass Stopcock, Class A, Serialized
Supplier: VWR International
76003-552 76003-554 76003-556 76003-558
76003-552CS 402.17 CAD
76003-552 76003-554 76003-556 76003-558
VWR® Burette, Glass Stopcock, Class A, Serialized
Burettes
VWR® Burettes are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 heat-resistant glass.
  • All burettes have a bore size of 2 mm
  • Calibrated to deliver (TD, EX) at 20°C
  • Calibrated to meet ASTM E 287 specifications
  • All burettes are calibrated according to Class A norms
  • An individual work certificate is included
VWR® Serialized Class A burettes are used to deliver solution in precisely measured, variable volumes. They are used primarily for titration, to deliver one reactant until the precise endpoint is reached. Calibration and tolerancesMore Product Information