YMC-Triart Diol-HILIC Hybrid Silica HPLC Columns, Diol-HILIC, 3 µm, YMC America
Supplier: YMC America,Inc
TDH12S03-0502WT TDH12S03-1002WT TDH12S03-1502WT TDH12S03-0503WT TDH12S03-1003WT TDH12S03-1503WT TDH12S03-1046WT TDH12S03-1546WT TDH12S03-0202WT TDH12S03-0302WT TDH12S03-0546WT TDH12S03-2546WT TDH12S03-H546WT TDH12S03-L502WT TDH12S03-L503WT TDH12S03-L546WT TDH12S03-05Q1PTP TDH12S03-10Q1PTP TDH12S03-15Q1PTP TDH12S03-0546PTP TDH12S03-1046PTP TDH12S03-1546PTP
89238-154EA 1466.08 CAD
89238-154 89238-156 89238-158 89238-160 89238-162 89238-164 89238-166 89238-168 10842-136 10842-144 10842-146 10842-148 10842-150 10842-152 10842-154 10842-156 75995-214 75995-216 75995-218 75995-220 75995-222 75995-224
YMC-Triart Diol-HILIC Hybrid Silica HPLC Columns, Diol-HILIC, 3 µm, YMC America
Chromatography Columns
Triart Diol-HILIC is a dihydroxypropane moiety bonded to an organic/inorganic hybrid based silica particle.
  • Dihydroxypropyl phase bonded to organic/inorganic hybrid silica particle
  • Ideal for the separation of polar and hydrophilic compounds that are not retained by reversed phase chromatography
  • Superior chemical durability
  • Identical selectivity over particle sizes
  • Ideal for HPLC
YMC-Triart Diol HILIC can retain extremely polar compounds while using eluents and buffers suitable for LC-MS.
The resulting product has excellent durability and usability across the wide pH range of pH 2 to 10. Triart Diol-HILIC is ideal for a separation of polar and hydrophilic compounds which are not retained by reversed phase (C18, C8, and others) chromatography.