YMC-Triart C18 Hybrid Silica HPLC Columns, C18, 3 µm, YMC America
Supplier: YMC America
YMC-Triart C18
TA12S03-0502WT TA12S03-3046WT TA12S03-H501WT TA12S03-15Q1PTP TA12S03-10Q1PTP TA12S03-L502WT TA12S03-0510WT TA12S03-05Q1PTP TA12S03-05P5WT TA12S03-10P5WT TA12S03-1503WT TA12S03-0302WT TA12S03-0546PTP TA12S03-3004WT TA12S03-1546WT TA12S03-1502WT TA12S03-1002WT TA12S03-1003WT TA12S03-1046PTP TA12S03-1504WT TA12S03-0202WT TA12S03-L506WT TA12S03-2546WT TA12S03-1546PTP TA12S03-1006WT TA12S03-L503WT TA12S03-0504WT TA12S03-2502WT TA12S03-H546WT TA12S03-1046WT TA12S03-1001WT TA12S03-0501WT TA12S03-R504WT TA12S03-H502WT TA12S03-2504WT TA12S03-L546WT TA12S03-0506WT TA12S03-1004WT TA12S03-0503WT TA12S03-0546WT TA12S03-2503WT TA12S03-H5P5WT
97049-002EA 2261.01 CAD
97049-002 10842-018 89216-994 76012-990 89216-992 97049-000 76012-992 76012-994 10841-672 76012-996 10841-670 10841-654 76012-998 76012-912 10841-674 97048-998 97048-984 10841-668 10842-002 10841-662 10841-660 97048-988 10841-666 76012-986 97048-986 10841-664 76012-988 10841-658 76012-914 97048-996 76012-916 10841-656 97048-994 97048-992 97048-990 10842-010 10842-012 97049-006 10842-014 97049-008 89216-998 10842-016
YMC-Triart C18 Hybrid Silica HPLC Columns, C18, 3 µm, YMC America
Chromatography Columns
YMC-Triart C18, 3 µm is a novel, organic/inorganic hybrid silica particle derivatized with C18 (L1 USP designation).
  • Chemical stability of the material combines with inherent mechanical durability of particles to contribute to long column lifetime
  • Available in columns packed with 3 µm particles, operating over a wide pH range of 1 to 12; this enables the manipulation of analytical retention and preparative loading
  • Proprietary end capping makes for excellent peak shapes of all types of compounds
  • Rigorous manufacturing controls result in legendary YMC column-to-column and lot-to-lot reproducibility
  • Unlike other hybrid materials, Triart C18, 3 µm exhibits a balanced hydrophobicity; it is more retentive than other hybrid materials, potentially simplifying the separation of 'difficult' peaks
  • Available in packed columns of various dimensions
  • May be operated in 100% aqueous mobile phases
The core material for all of the YMC-Triart products, the particle, is inspired by YMC's flow chemistry technology. Inherent in YMC’s process is a high level of temperature and particle formation control. As a result, Triart particles are consistent, reproducible, and scalable across many particle sizes. The hybrid Triart particles produce a material that is highly resilient and robust when used at extreme pH and elevated temperature. In addition, these particles are very stable under mechanical stress, such as high pressure. YMC-Triart C18, 3 µm can be used at temperatures up to 70 °C at pH 1 to 7, and 50 °C from pH 7 to 12. This allows the chromatographer to use temperature and pH as tools to optimize suitable separation conditions, and be less worried about the limitations of stationary phase ruggedness.
Triart C18 is enhanced by an end capping process that eliminates the secondary interactions that contribute to poor peak shape, and is ideal for analysis of all types of compounds. Triart C18 is an ideal first-choice method development column.